Spatulas, Trulla type
175 mm
Katalognr. 231-0393
Anbieter: NICKEL ELECTRO
Direkt zu
Product Details & Documents
Stainless steel 18/8 non-magnetic, bright finish. Double ended spatulas with trowel shaped scoops for handling large quantities of material.
- Excellent thermal resistance, autoclavable
- Excellent chemical resistance
- Quick and easy to clean
Technische Daten
- Länge:175 mm