Protective visor, anti-COVID face shield
Katalognr. 111-2468
Anbieter: MULTIROIR
Technische Daten
- Bezeichnung:Protective visor, black
- Seq:01
- VE:10
Technische Daten
Über diesen Artikel
Anti-COVID personal protective visors are adjustable and comfortable, allowing for extended use. The ergonomics of the head circumference allow for convenient assembly and adjustment.
- Facial protection from droplets, saliva, splash, oil, and dust
- Head circumference is polypropylene material
- Visors are made from transparent, 240 micrometer, PET (polyethylene terephthalate) material
- Transparency guaranteed
- Suitable for use with glasses or protective masks
- Universal size (adjustable)
- Delivered unmounted, easy assembly
These protective visors have been designed to ensure employee safety and security. Employees required to provide direct contact with customers can be guaranteed protection against respiratory viruses. The visors can be used in a wide variety of activities, and are compatible with glasses or protective face masks.