Safety spectacles, Contemporary 573 Diamond, medium
Catalog # 111-2463
Supplier: UNIVET
Specifications
- Typ:Anti-UV, anti-scratch, anti-fog
- Farbe:Dunkelgrau
- Verglasung:Clear PC
- VE:1
Specifications
About this item
Contemporary series combines elegance and style in proetction, changing the concept of safety glasses.
- Light and extremely tough
- Integrated side shields
- PA12 nylon frame with zero metallic components
- Glossy varnish with shaded effect
- Clear KN lenses, with anti-fog and anti-scratch ultra performing Vanguard PLUS treatment
Elegant and graceful for women, dynamic for men. Glasses can be employed for prolonged use as they do not create distorsions and allow a correct color perception, sensibly reducing visual fatigue.
Suitable for mechanical work in good visibility conditions.
Size: 53
Certifications: EN-166, EN-170.