ContactUsLinkComponent
Molybdate reagent solution, for photometric phosphate determination acc. to DIN 38405-D11-1, Supelco®
Molybdate reagent solution, for photometric phosphate determination acc. to DIN 38405-D11-1, Supelco®
Catalog # 69888-100ML
Supplier:  Merck
CAS Number:  
Molybdate reagent solution, for photometric phosphate determination acc. to DIN 38405-D11-1, Supelco®
Catalog # 69888-100ML
Supplier:  Merck
CAS Number:  
Specifications
  • VE:
    100 mL
  • Bezeichnung:
    Molybdate reagent solution
Specifications
About this item
Solution contains hexaammonium heptamolybdate ((NH₄)₆Mo₇O₂₄.4H₂O); sulfuric acid (H₂SO₄); potassium antimony(III) oxide tartrate(K(SbO)C₄H₄O₆.0.5H₂O);water
Caution: Solution is corrosive (GHS05) please consult MSDS.