SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standards
Organische Standards
Testosterone enanthate, analytical standard, Supelco®
Testosterone enanthate, analytical standard, Supelco®
T3006-1G
:
Merck
:
Testosterone enanthate, analytical standard, Supelco®
T3006-1G
:
Merck
:
VE:
1 g
Normen:
analytical standard
CAS Nr.:
315-37-7
Bezeichnung:
Testosterone enanthate
Bezeichnung:
Testosterone enanthate