Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Supelco® TraceCERT® Organic Reference Standard, Benzene solution

Supelco® TraceCERT® Organic Reference Standard, Benzene solution

Benzene solution, 200 μg/ml in methanol, Certified reference material, TraceCERT®, 1
Katalognr. CRM48617
Anbieter:   Merck
Supelco® TraceCERT® Organic Reference Standard, Benzene solution
Katalognr. CRM48617
Anbieter:  Merck
CAS Nummer:  
Supelco® TraceCERT® Organic Reference Standard, Benzene solution

Benzene solution, 200 μg/ml in methanol, Certified reference material, TraceCERT®, 1

CAS Nummer:  
Preis
SFr. 28.10
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
  • GHS signal: GHS02, GHS06, GHS08

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.

Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311

Documents

Technische Daten

Technische Daten

  • Konzentration:
    200 μg/ml in methanol
  • VE:
    1
  • Normen:
    Certified reference material, TraceCERT®
  • Bezeichnung:
    Benzene solution
  • Cat. No.:
    CRM48617
  • CAS Nr.:
    71-43-2
  • Molekulargewicht:
    78,11
  • Beschreibung Gefahrenklasse:
    H225 - H301 + H311 + H331 - H370
  • Flammpunkt:
    9,7 °C
  • Lagerbedingungen:
    2...30 °C
  • Shipping conditions:
    STD
  • M-Clarity Qualitätsstufe:
    MQ100

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.