Combined six cation standard-II, Dionex™
DION046070
: DIONEX
:
- VE:50 mL
- Bezeichnung:Dionex™ combined six cation standard-II
- Environmentally Preferable:
Ion standards are used for a variety of routine anion, cation, and standard determinations.
Increase the productivity and reduce standard preparation errors with Dionex™ ion standards for ion chromatography. Available for routine anion and cation determinations, these single and combined ion standards are made with high purity, reagent-grade chemicals and water.
: Traceable to NIST.