ContactUsLinkComponent
Bucket system, 20 L
Bucket system, 20 L
Catalog # CONT2769-KIT
Supplier:  CONTEC
undefined
Bucket system, 20 L
Catalog # CONT2769-KIT
Supplier:  CONTEC
Specifications
  • Bezeichnung:
    Double bucket system,two Buckets, cart with heavy-duty casters
  • Verpackt:
    1/case
  • VE:
    1 KIT
Specifications
About this item
Polypropylene bucket system is a stainless steel rolling cart with two polypropylene 20 L buckets, stainless steel drip cap, and angled stainless steel wringer.
  • The red and grey double bucket system allows for differentiation between cleaning and rinsing solutions
  • Compatible with all Contec® flat mops, including our PREMIRA™ advanced environmental hygiene system and Klean Max™
Durable, yet lightweight construction with four standard casters makes it easy to manoeuvre around pharmacy cleanrooms.