Natriumammoniumhydrogenphosphat Tetrahydrat ≥99,0% (durch nicht wässrige Titration), Puriss. p.a., Fluka™
Catalog # 71283-1KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Packung:Plastic bottle
- VE:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:99.0 - 101.0 %
- Silver (Ag):Max. 5 ppm
- Aluminium (Al):Max. 5 ppm
- Arsenic (As):Max. 0.5 ppm
- Barium (Ba):Max. 5 ppm
- Bismuth (Bi):Max. 5 ppm
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 10 ppm
- Lithium (Li):Max. 5 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Molybdenum (Mo):Max. 5 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Strontium (Sr):Max. 5 ppm
- Thallium (Tl):Max. 10 ppm
- Zinc (Zn):Max. 5 ppm
- Total S (as SO₄):Max. 50 ppm
- Chloride (Cl):Max. 10 ppm
Specification Test Results
About this item
:
:209.07 g/mol
Density:1,544 g/cm³
Storage Temperature:Raumtemperatur
CAS Number:7783-13-3
Formula:NaNH₄HPO₄·4H₂O
MDL Number:MFCD00066970