Springen zu Kopfbereich
Springen zu Hauptinhalt
Springen zu Fußzeile
Erweiterte Suche
Erweiterte Suche
Sicherheitsdatenblatt (SDS)
Zertifikate
Herstellerdaten
0
SFr. 0.00
Order Entry
Login
/
Registrieren
Products
Services
Digital Solutions
Applications
Support
About
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
Switzerland
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
ContactUsLinkComponent
Heim
Standards
Umweltanalyse-Standards
Chlorinated pesticides - additional mix, (CRM), Supelco®
Chlorinated pesticides - additional mix, (CRM), Supelco®
Chlorinated pesticides - additional mix, (CRM), Supelco®
Catalog # PE1481-2ML
Supplier:
Merck
CAS Number:
Expand image
Chlorinated pesticides - additional mix, (CRM), Supelco®
Catalog # PE1481-2ML
Supplier:
Merck
CAS Number:
Specifications
VE:
2 mL
Bezeichnung:
Chlorinated Pesticides - Additional Mix
Environmentally Preferable:
Specifications
About this item
Product Details
Product Details