Ethidium monoazide bromide (EMA) DNA/RNA Vergleichsfarbstoff
Catalog # 40015.
Supplier: Biotium
CAS Number:
Specifications
- VE:5 mg
- Seq:2210
Specifications
About this item
:
:420.305 g/mol
Storage Temperature:Kühlschrank
CAS Number:58880-05-0
Formula:C₂₁H₁₈BrN₅