Safety spectacles, Polarvision
Katalognr. UVEX9231.960
Anbieter: UVEX
Technische Daten
- Typ:Anti-UV, anti-scratch
- Farbe:Schwarz/weiß
- Verglasung:Grey PC
- VE:1
Technische Daten
Über diesen Artikel
Comfortable safety spectacles with polarisation filter.
- Polarisation filter protects against reflections and glare (e.g. water surfaces and highly polished materials)
- Scratch-resistant coated
Suitable for workstations with reflective surfaces (offshore, maritime, drivers, construction industry).