Tube rack, neoRack®
Catalog # NEOL240121634
Supplier: NEOLAB
Specifications
- Bohrung Ø:30 mm
- Anz. Bohrungen:4×4
- Farbe:Rot
- Höhe:70 mm
- L×B:146×146 mm
- Material:PP
- VE:1
Specifications
About this item
This tube rack is open at the front and are pierced by three frame levels or square grids, which are inserted and latched into the side and back sections.
- Alphanumeric identification of the openings
- Stackable
- Colour coded
- Can be quickly assembled and disassembled
- Can easily be stowed in a cupboard or drawer
- Plugged together blue/red
Made of durable polypropylene, temperature resistant and autoclavable from 90 to +120 °C.
The sides and back as well as the bottom floor are blue and the two upper levels to fix the tubes are colour coded.