ContactUsLinkComponent
Micro BCA protein assay kit
  BSBTAR1110
 :  
undefined
Micro BCA protein assay kit
  BSBTAR1110
 :  
  • VE:
    1 KIT
  • Bezeichnung:
    Micro BCA Protein Assay Kit, sufficient for 200 tube assays or 1300 microplate assays
The Micro BCA Protein Assay Kit is a ready to use, detergent-compatible, Western blot related, total protein analysis reagent used for the quick determination of total protein concentration of dilute protein solutions by measuring absorbance at 562 nm and comparing to a protein standard absorbance vs. concentration curve.
  • Light blue liquid
  • Active in alkaline medium
  • Stable at room temperature for one year
The Micro BCA Protein Assay Kit provides an ultra-sensitive method to detect and quantify protein concentration. The kit has been optimized for determining total protein content in low protein concentration samples (0.5 to 20 μg/ml). The kit is sensitive to copper reductant and chelator instead of to both ionic and non-ionic detergents.
 : The kit contains micro BCA reagent A (1×100 ml), micro BCA reagent B (1×100 ml), micro BCA reagent C (1×5 ml) and albumin (BSA) standards (1×20 ml) (2 mg/ml).