AuftragsverfolgungKontakt
HPLC columns, NUCLEOSHELL® PFP
Supplier:   MACHEREY-NAGEL
HPLC columns, NUCLEOSHELL® PFP
Supplier:  MACHEREY-NAGEL
Certificates
HPLC columns, NUCLEOSHELL® PFP
14 Options Available Below
Product Details & Documents
NUCLEOSHELL® PFP based on core-shell particle technology for fast and efficient HPLC.
  • Efficient core-shell silica gel
  • Pentafluorophenyl propyl (PFP) modification
  • Hydrophobic phase with alternative selectivity in comparison to classical C18 modifications
The separation principle based on four retention mechanisms such as polar interactions, dipole-dipole interactions, π-π interactions, hydrophobic interactions.
Suitable for the separation of polar compounds like pharmaceuticals such as antibiotics, aromatic and unsaturated compounds, phenols, halogenated hydrocarbons and isomers. It shows a high retention of basic compounds.
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).
Specifications for Product Family
Specifications
  • Catalog No:
  • MANA763532.20
  • MANA763534.20
  • MANA763536.20
  • MANA763532.30
  • MANA763534.30
  • MANA763536.30
  • MANA763532.40
  • MANA763534.40
  • MANA763536.40
  • MANA763532.46
  • MANA763534.46
  • MANA763536.46
  • MANA763538.20
  • MANA763538.30
  • Kohlenstoffbelastung:
  • ~3,0%
  • ~3,0%
  • ~3,0%
  • ~3,0%
  • ~3,0%
  • ~3,0%
  • ~3,0%
  • ~3,0%
  • ~3,0%
  • ~3,0%
  • ~3,0%
  • ~3,0%
  • ~3,0%
  • ~3,0%
  • Endcapped:
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • Ja
  • pH-Bereich:
  • 1 - 9
  • 1 - 9
  • 1 - 9
  • 1 - 9
  • 1 - 9
  • 1 - 9
  • 1 - 9
  • 1 - 9
  • 1 - 9
  • 1 - 9
  • 1 - 9
  • 1 - 9
  • 1 - 9
  • 1 - 9
Specifications
Product Family Options
Analytical columns
Product Information
  • Säulenform
    Phase
    Porengröße
    Partikelgröße
    Ø int.
    Länge
    Availability
    Price
  • Analytisch
    PFP
    90 Å
    2,7 µm
    2,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 817.00
    SFr. 817.00
    1 * 1 ST
       SFr. 817.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763532.20
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
  • Analytisch
    PFP
    90 Å
    2,7 µm
    2,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 923.00
    SFr. 923.00
    1 * 1 ST
       SFr. 923.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763534.20
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
  • Analytisch
    PFP
    90 Å
    2,7 µm
    2,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 923.00
    SFr. 923.00
    1 * 1 ST
       SFr. 923.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763536.20
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
  • Analytisch
    PFP
    90 Å
    2,7 µm
    3,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 817.00
    SFr. 817.00
    1 * 1 ST
       SFr. 817.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763532.30
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
  • Analytisch
    PFP
    90 Å
    2,7 µm
    3,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 934.00
    SFr. 934.00
    1 * 1 ST
       SFr. 934.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763534.30
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
  • Analytisch
    PFP
    90 Å
    2,7 µm
    3,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 934.00
    SFr. 934.00
    1 * 1 ST
       SFr. 934.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763536.30
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
  • Analytisch
    PFP
    90 Å
    2,7 µm
    4,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 817.00
    SFr. 817.00
    1 * 1 ST
       SFr. 817.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763532.40
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
  • Analytisch
    PFP
    90 Å
    2,7 µm
    4,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 934.00
    SFr. 934.00
    1 * 1 ST
       SFr. 934.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763534.40
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
  • Analytisch
    PFP
    90 Å
    2,7 µm
    4,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 934.00
    SFr. 934.00
    1 * 1 ST
       SFr. 934.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763536.40
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
  • Analytisch
    PFP
    90 Å
    2,7 µm
    4,60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 890.00
    SFr. 890.00
    1 * 1 ST
       SFr. 890.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763532.46
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
  • Analytisch
    PFP
    90 Å
    2,7 µm
    4,60 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’030.00
    SFr. 1’030.00
    1 * 1 ST
       SFr. 1’030.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763534.46
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
  • Analytisch
    PFP
    90 Å
    2,7 µm
    4,60 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’030.00
    SFr. 1’030.00
    1 * 1 ST
       SFr. 1’030.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763536.46
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
Guard columns
Product Information
  • Säulenform
    Phase
    Porengröße
    Partikelgröße
    Ø int.
    Länge
    Availability
    Price
  • Vorsäule
    PFP
    90 Å
    2,7 µm
    2,00 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - SFr. 179.00
    SFr. 179.00
    1 * 3 ST
       SFr. 179.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763538.20
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9
  • Vorsäule
    PFP
    90 Å
    2,7 µm
    3,00 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - SFr. 179.00
    SFr. 179.00
    1 * 3 ST
       SFr. 179.00
    Specifications:
    More
    Specifications
    Cat. No.
    MANA763538.30
    Kohlenstoffbelastung
    ~3,0%
    Endcapped
    Ja
    pH-Bereich
    1 - 9