AuftragsverfolgungKontakt
Super Safe sampler | Avantor
Super Safe sampler
Sampler for bioreactors
Katalognr. INFO65373
Anbieter:   INFORS
Super Safe sampler
Katalognr. INFO65373
Anbieter:  INFORS
Super Safe sampler
Sampler for bioreactors
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
This sampler allows you to take smallest samples without wasting any culture.
  • Sterile sampling without laminar flow hood
  • Truly aseptic and prevents faulty operation and contamination
  • More precise results especially in small scale bioprocesses – especially important for small culture vessels and parallel bioprocess
Can be used for all types of small culture vessels like bags, spinner flasks, stirred tank reactors.
Technische Daten
  • Bezeichnung:
    Sampler for bioreactors