LED illuminator
LED illuminator
Katalognr. ZEIS4239049902000
Anbieter: ZEISS
Direkt zu
Product Details & Documents
LED illuminator for transmitted light for direct attachment to the condenser carrier.
Contains sliders with diffusion disk with 10° divergence angle for visual observation and with 80° divergence angle for documentation. Slider can also be used as shutter against interfering autofluorescence of the LEDs in epi-fluorescence applications.
Technische Daten
- Bezeichnung:LED illuminator