Human Recombinant BAFF (from E. coli)
Über diesen Artikel
B cell-activating factor (BAFF), or B lymphocyte stimulator (BLyS), is a type II member of the tumor necrosis factor (TNF) superfamily. BAFF is expressed as a transmembrane protein on T cells, macrophages, and dendritic cells. The transmembrane domain of BAFF can also be cleaved to produce a soluble protein fragment.
- High quality
- Low endotoxin
- Affordable
BAFF binds to the TNF receptors known as B cell maturation antigen (BCMA), transmembrane activator and CAML interactor (TACI), and BAFF receptor (BAFFR). BAFF is important for the survival and maturation of peripheral B cells. Human BAFF shows activity on mouse splenocytes.
: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
: For research use only.
4 Varianten
Product Details & Documents
B cell-activating factor (BAFF), or B lymphocyte stimulator (BLyS), is a type II member of the tumor necrosis factor (TNF) superfamily. BAFF is expressed as a transmembrane protein on T cells, macrophages, and dendritic cells. The transmembrane domain of BAFF can also be cleaved to produce a soluble protein fragment.
- High quality
- Low endotoxin
- Affordable
BAFF binds to the TNF receptors known as B cell maturation antigen (BCMA), transmembrane activator and CAML interactor (TACI), and BAFF receptor (BAFFR). BAFF is important for the survival and maturation of peripheral B cells. Human BAFF shows activity on mouse splenocytes.
: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
: For research use only.
Spezifikationen für diese Produktfamilie
Technische Daten
Technische Daten
- Catalog No:
- SHBT100-24-5UG
- SHBT100-24-100UG
- SHBT100-24-500UG
- SHBT100-24-1MG
- Protein/Peptid-Name:
- BAFF
- BAFF
- BAFF
- BAFF
- Proteinsynonym:
- TALL|THANK|BLyS
- TALL|THANK|BLyS
- TALL|THANK|BLyS
- TALL|THANK|BLyS
- Protein/Peptid-Typ:
- Rekombinant
- Rekombinant
- Rekombinant
- Rekombinant
- Protein-Funktion:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Spezies:
- Human
- Human
- Human
- Human
- Ursprung:
- E. coli
- E. coli
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gen-ID:
- Q9Y275
- Q9Y275
- Q9Y275
- Q9Y275
- Sequence:
- MHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLL
- MHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLL
- MHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLL
- MHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLL
- Frei von Proteasen:
- N
- N
- N
- N
- Tierfrei:
- N
- N
- N
- N
- Frei von Nukleasen:
- N
- N
- N
- N
- Frei von Trägern:
- Y
- Y
- Y
- Y
- Frei von Endotoxinen:
- N
- N
- N
- N
- Endotoxinspiegel:
- Low
- Low
- Low
- Low
- Endotoxingehalt:
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- UniProtKB:
- Q9Y275
- Q9Y275
- Q9Y275
- Q9Y275
- Biologische Aktivität:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥90%
- ≥90%
- ≥90%
- ≥90%
- Sorte:
- RUO
- RUO
- RUO
- RUO
- Zusammensetzung:
- 10 mM sodium phosphate, pH 7,5
- 10 mM sodium phosphate, pH 7,5
- 10 mM sodium phosphate, pH 7,5
- 10 mM sodium phosphate, pH 7,5
- Molekulargewicht:
- 18,5 kDa
- 18,5 kDa
- 18,5 kDa
- 18,5 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Lagerbedingungen:
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- Transporttemperatur:
- RT
- RT
- RT
- RT
Technische Daten
Product Family Options
Produktinformationen
- PackungVEVerfügbarkeitPreis
- Spezifikationen:
Mehr
Specifications
Cat. No.SHBT100-24-5UGProtein/Peptid-NameBAFFProteinsynonymTALL|THANK|BLySProtein/Peptid-TypRekombinantProtein-FunktionCytokineSpeziesHumanUrsprungE. coliKonjugationUnconjugatedGen-IDQ9Y275SequenceMHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLLFrei von ProteasenNTierfreiNFrei von NukleasenNFrei von TrägernYFrei von EndotoxinenNEndotoxinspiegelLowEndotoxingehaltEndotoxin LAL (EU/µg) ≤1UniProtKBQ9Y275Biologische AktivitätBioactivity: No biological activity data is available at this timePurity≥90%SorteRUOZusammensetzung10 mM sodium phosphate, pH 7,5Molekulargewicht18,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlLagerbedingungen−20 °CTransporttemperaturRT - Spezifikationen:
Mehr
Specifications
Cat. No.SHBT100-24-100UGProtein/Peptid-NameBAFFProteinsynonymTALL|THANK|BLySProtein/Peptid-TypRekombinantProtein-FunktionCytokineSpeziesHumanUrsprungE. coliKonjugationUnconjugatedGen-IDQ9Y275SequenceMHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLLFrei von ProteasenNTierfreiNFrei von NukleasenNFrei von TrägernYFrei von EndotoxinenNEndotoxinspiegelLowEndotoxingehaltEndotoxin LAL (EU/µg) ≤1UniProtKBQ9Y275Biologische AktivitätBioactivity: No biological activity data is available at this timePurity≥90%SorteRUOZusammensetzung10 mM sodium phosphate, pH 7,5Molekulargewicht18,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlLagerbedingungen−20 °CTransporttemperaturRT - Spezifikationen:
Mehr
Specifications
Cat. No.SHBT100-24-500UGProtein/Peptid-NameBAFFProteinsynonymTALL|THANK|BLySProtein/Peptid-TypRekombinantProtein-FunktionCytokineSpeziesHumanUrsprungE. coliKonjugationUnconjugatedGen-IDQ9Y275SequenceMHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLLFrei von ProteasenNTierfreiNFrei von NukleasenNFrei von TrägernYFrei von EndotoxinenNEndotoxinspiegelLowEndotoxingehaltEndotoxin LAL (EU/µg) ≤1UniProtKBQ9Y275Biologische AktivitätBioactivity: No biological activity data is available at this timePurity≥90%SorteRUOZusammensetzung10 mM sodium phosphate, pH 7,5Molekulargewicht18,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlLagerbedingungen−20 °CTransporttemperaturRT - Spezifikationen:
Mehr
Specifications
Cat. No.SHBT100-24-1MGProtein/Peptid-NameBAFFProteinsynonymTALL|THANK|BLySProtein/Peptid-TypRekombinantProtein-FunktionCytokineSpeziesHumanUrsprungE. coliKonjugationUnconjugatedGen-IDQ9Y275SequenceMHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLLFrei von ProteasenNTierfreiNFrei von NukleasenNFrei von TrägernYFrei von EndotoxinenNEndotoxinspiegelLowEndotoxingehaltEndotoxin LAL (EU/µg) ≤1UniProtKBQ9Y275Biologische AktivitätBioactivity: No biological activity data is available at this timePurity≥90%SorteRUOZusammensetzung10 mM sodium phosphate, pH 7,5Molekulargewicht18,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlLagerbedingungen−20 °CTransporttemperaturRT