QUANTOFIX® Test Strips, Swimming Pool Test
Über diesen Artikel
Semi-qualitative test strips for water qualities in swimming pool.
- Easy dip-and-read procedure Multi-stage colour scale Provides reliable results within 30 seconds No accessories necessary for evaluation Desiccant in the stopper of tube container for optimal protection of the strips against humidity
These strips are used for quick and easy determination of water qualities in swimming pool. The test provides valuable results about the water hardness, the total alkalinity, free and total chlorine and pH.
2 Varianten
Product Details & Documents
Semi-qualitative test strips for water qualities in swimming pool.
- Easy dip-and-read procedure Multi-stage colour scale Provides reliable results within 30 seconds No accessories necessary for evaluation Desiccant in the stopper of tube container for optimal protection of the strips against humidity
These strips are used for quick and easy determination of water qualities in swimming pool. The test provides valuable results about the water hardness, the total alkalinity, free and total chlorine and pH.
Dokumente
Dip and Read Tests, Rapid Tests.pdf
Weitere Produktdetails
SDS_Swimming Pool Test SDS_Swimming Pool Test 5 in 1 (50 Strips)
Sicherheitsdatenblatt (SDS)
Spezifikationen für diese Produktfamilie
Technische Daten
Technische Daten
- Catalog No:
- MANA90752
- MANA90759
- Parameter:
- All reagents included
- All reagents included
- Konformität:
- Not CE certified and no hazardous material
- Not CE certified and no hazardous material
- Inhalt:
- 50 test strips in a tube
- 50 test strips in a tube
- Instrument:
- No
- No
- Platform:
- Semi-quantitative test strips
- Semi-quantitative test strips
- Haltbarkeit (Monate):
- 24 (from production)
- 24 (from production)
- Gewicht:
- 34,1 g
- 35,5 g
Technische Daten
Product Family Options
Produktinformationen
- BezeichnungMessbereichChemische MethodeAbmessungenTests pro KitVEVerfügbarkeitPreis
- Swimming pool test 3 in 10 • 0.5 • 1 • 3 • 5 • 10 - mg/L - Cl₂ ; 0 • 80 • 120 • 180 • 240 - mg/L - CaCO₃ ; 6.4 • 6.8 • 7.2 • 7.6 • 8.4Free chlorine/ total chlorine: Colour reaction with azines; Alcalinity: Reaction with an acid and mixture of pH indicators; pH: Colour reaction of a pH indicator40×40×120 mm (package)50 test(s)50Einloggen für Verfügbarkeit1 * 50 STSFr. 44.70Spezifikationen:
Mehr
Specifications
Cat. No.MANA90752ParameterAll reagents includedKonformitätNot CE certified and no hazardous materialInhalt50 test strips in a tubeInstrumentNoPlatformSemi-quantitative test stripsHaltbarkeit (Monate)24 (from production)Gewicht34,1 g - Swimming pool test 5 in 10 • 0.5 • 1 • 3 • 5 • 10 - mg/L - Cl₂ ; 0 • 80 • 120 • 180 • 240 - mg/L - CaCO₃ ; 6.4 • 6.8 • 7.5 • 7.6 • 8.4 ; 0 • 1 • 3 • 5 • 10 - mg/L - Cl₂Alcalinity: Reaction with an acid and mixture of pH indicators; Free chlorine/ total chlorine: Colour reaction with azines; pH: Colour reaction of a pH indicator; Total hardness: Colour reaction with complex indicator40×40×120 mm (package)50 test(s)1Einloggen für Verfügbarkeit1 * 1 STSFr. 36.40Spezifikationen:
Mehr
Specifications
Cat. No.MANA90759ParameterAll reagents includedKonformitätNot CE certified and no hazardous materialInhalt50 test strips in a tubeInstrumentNoPlatformSemi-quantitative test stripsHaltbarkeit (Monate)24 (from production)Gewicht35,5 g