
4-Methylpyridazin
Catalog # MOLE29354234-100MG
Supplier: Molekula
CAS Number:
Specifications
- Packung:Glass bottle for solids
- VE:100 mg
- Seq:2440
Specifications
About this item
:
Boiling Pt:66 °C (0.8 mmHg)
Density:1,0641 g/cm³ (20 °C)
:94.12 g/mol
:GHS07
CAS Number:1120-88-3
Formula:C₅H₆N₂
MDL Number:MFCD00011594
Boiling Pt:66 °C (0,8 mmHg)