Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Imidazol

Imidazol

Corrosive
Health
Irritant
Supplier:   EUROCLONE
Imidazol
Supplier:  EUROCLONE
Certificates
Imidazol

4 Options Available Below

Product Details & Documents
Synonyms:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS Number:288-32-4
  • Formula:C₃H₄N₂
  • MDL Number:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Merck Index:13,04935
  • Boiling Pt:255 °C (1013 hPa)
  • Melting Pt:89…90 °C
  • Density:1,18 g/cm³ (20 °C)
  • Flash Pt:135 °C
  • Storage Temperature:Raumtemperatur
  • EINECS:206-019-2
  • UN:2923
  • ADR:8,III
Product Family Options

Product Information

  • Packung
    VE
    Availability
    Price
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.