Cetrimoniumbromid
Catalog # EURLEMR288500
Supplier: EUROCLONE
CAS Number:
Specifications
- VE:1 KIT
- Seq:0075
Specifications
About this item
:
:364.45 g/mol
Merck Index:13,02034
Melting Pt:237…243 °C
Flash Pt:244 °C (closed cup)
Storage Temperature:Raumtemperatur
EINECS:200-311-3
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS05
:GHS09
CAS Number:57-09-0
Formula:CH₃(CH₂)₁₅N(Br)(CH₃)₃
MDL Number:MFCD00011772