Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Kaliumsulfat

Kaliumsulfat

Supplier:   EUROCLONE
Kaliumsulfat
Supplier:  EUROCLONE
Certificates

2 Options Available Below

Product Details & Documents
  • CAS Number:7778-80-5
  • Formula:K₂SO₄
  • MDL Number:MFCD00011388
  • Molecular Weight:174.26 g/mol
  • Merck Index:13,07759
  • Boiling Pt:1698 °C (1013 hPa)
  • Melting Pt:1069 °C
  • Density:2,66 g/cm³ (20 °C)
  • Storage Temperature:Raumtemperatur
  • EINECS:231-915-5
Product Family Options

Product Information

  • Packung
    VE
    Availability
    Price
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.