Order Entry
Switzerland
ContactUsLinkComponent
2,4,6-Trichlorbiphenyl (PCB Nr. 30)
2,4,6-Trichlorbiphenyl (PCB Nr. 30)
Catalog # PROOCIL-PCB-30-C
CAS Number:  
2,4,6-Trichlorbiphenyl (PCB Nr. 30)
Catalog # PROOCIL-PCB-30-C
CAS Number:  

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :257.55 g/mol
UN:2315
ADR:9,II
CAS Number:35693-92-6
Formula:C₁₂H₇Cl₃