Order Entry
Switzerland
ContactUsLinkComponent
Formaldehyd 4% in wässriger Lösung, EM Grad
Irritant
Health
Formaldehyd 4% in wässriger Lösung, EM Grad
Catalog # ELMI157-4
CAS Number:  
Irritant
Health
Formaldehyd 4% in wässriger Lösung, EM Grad
Catalog # ELMI157-4
CAS Number:  

Specifications

  • Packung:
    Glass ampoule
  • VE:
    1
  • Seq:
    2883
  • Pk Info:
    10×10 ml

Specifications

About this item

 :
 :30.03 g/mol
Storage Temperature:Raumtemperatur
UN:2209
ADR:8,III
 :GHS07
 :GHS08
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274