Order Entry
Switzerland
ContactUsLinkComponent
2,4,6-Tri-(dimethylaminomethyl)phenol
  ELMI13600
 :  
2,4,6-Tri-(dimethylaminomethyl)phenol
  ELMI13600
 :  

 

  • Packung:
    Bottle
  • VE:
    25 mL
  • Seq:
    2315
  • Environmentally Preferable:

 

 

  • Refractive Index at 25 °C:
    1.514
  • Specific Gravity at 25 °C:
    0.973

 

 

 : Tris-2,6-dimethylaminomethylphenol, 2,6-Tris-N,N-dimethylaminomethylfenol, DMP-30, Epon® DMP-30 substitute
 :
 :265.4 g/mol
 :2735
 :GHS07
 :Raumtemperatur
 :90-72-2
 :C₁₅H₂₇N₃O
 :MFCD00008330
 :0.973 g/ml (25 °C)
 :8 III