AuftragsverfolgungKontakt
Phenoxyethanol
Glass bottle for solids, 1 kg
Catalog # MOLEM30650496
Irritant
Marketsource
Supplier:   Molekula
Phenoxyethanol
Catalog # MOLEM30650496
Supplier:  Molekula
Marketsource
CAS Number:  122-99-6
Phenoxyethanol
Phenoxyethanol
Glass bottle for solids, 1 kg
CAS Number:  122-99-6
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
  • CAS Number:122-99-6
  • Formula:C₆H₅OCH₂CH₂OH
  • MDL Number:MFCD00002857
  • Molecular Weight:138.17 g/mol
  • Merck Index:12,07410
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:13 °C
  • Density:1,1094 g/cm³ (20 °C)
  • Flash Pt:121 °C (geschlossener Tiegel)
  • Storage Temperature:Raumtemperatur
  • EINECS:204-589-7
Specifications
Specifications
  • Packung:
    Glass bottle for solids
  • VE:
    1 kg