Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Bisacrylamid

Bisacrylamid

Irritant
Supplier:   EUROCLONE
Bisacrylamid
Supplier:  EUROCLONE
Certificates
Bisacrylamid

3 Options Available Below

Product Details & Documents
Synonyms:  Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
  • CAS Number:110-26-9
  • Formula:(H₂C=CHCONH)₂CH₂
  • MDL Number:MFCD00008625
  • Molecular Weight:154.17 g/mol
  • Boiling Pt:125 °C (3,33 hPa)
  • Melting Pt:185 °C
  • Density:1,235 g/cm³ (20 °C)
  • Flash Pt:138 °C
  • Storage Temperature:Kühlschrank
  • EINECS:203-750-9
Product Family Options

Product Information

  • Packung
    VE
    Availability
    Price
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.