

Ethanol ≥96% (v/v), Puriss. p.a. ACS, Reag. Ph. Eur. analytisches Reagens, Riedel-de Haën™
Catalog # 32294-4X2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Packung:PE-Flasche
- VE:10 L
- Seq:5214
- Environmentally Preferable:Unbedenklicher für Mensch und Umwelt
- Umweltzertifizierung:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Pk Info:4x 2.5 L
Specifications
Specification Test Results
- Appearance:complying
- Identity A:complying
- Identity B:complying
- Assay:95.1 - 96.9 Vol.%
- Boiling range:78 - 79 °C
- Density (D 20/20):0.8050 - 0.8120
- Non-volatile matter:Max. 0.001 %
- Free acid (as CH₃COOH):Max. 0.003 %
- Free alkali (as NH₃):Max. 0.0001 %
- Cadmium (Cd):Max. 0.00001 %
- Copper (Cu):Max. 0.00001 %
- Iron (Fe):Max. 0.00001 %
- Nickel (Ni):Max. 0.00001 %
- Lead (Pb):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- Heavy metals (as Pb):Max. 0.0001 %
- KMnO₄ reducing matter:complying
- Acetone (GC):Max. 0.001 %
- Aldehydes (as CH₃CHO):Max. 0.0005 %
- Formaldehyde:Max. 0.0005 %
- Methanol (GC):Max. 100 Vol.ppm
- 2-propanol (GC):Max. 0.003 %
- Volatile impurities (GC):complying
- Reaction against H₂SO₄:complying
- Mixable with H₂O:complying
- UV-absorption:complying
- APHA:Max. 10
Specification Test Results
About this item
:
:46.07 g/mol
Boiling Pt:78…79 °C (1.013 hPa)
Melting Pt:–115 °C
Density:0.805…0.812 g/cm3 (20 °C)
Flash Pt:12 °C
Storage Temperature:Raumtemperatur
EINECS:200-578-6
UN:1170
ADR:3,II
:GHS07
:GHS02
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568