ContactUsLinkComponent
Flammable
Toxic
Corrosive
N-Ethyldiisopropylamin
Catalog # MOLEM28003285
Supplier:  Molekula
CAS Number:  
Flammable
Toxic
Corrosive
N-Ethyldiisopropylamin
Catalog # MOLEM28003285
Supplier:  Molekula
CAS Number:  
Specifications
  • Packung:
    Glasflasche
  • VE:
    100 mL
  • Seq:
    2440
Specifications
About this item
 :
 :129.25 g/mol
Boiling Pt:127 °C (1013 hPa)
Melting Pt:–50 °C
Density:0.76 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Raumtemperatur
EINECS:230-392-0
UN:2733
ADR:3,II
 :GHS02
 :GHS06
 :GHS05
CAS Number:7087-68-5
Formula:C₈H₁₉N
MDL Number:MFCD00008868