Stearinsäure
Anbieter: MATREYA
Direkt zu
Über diesen Artikel
Synonyme: Octadecanoic acid, Octadecylic acid
- CAS Nummer:57-11-4
- Formel:C₁₈H₃₆O₂
- MDL-Nummer:MFCD00002752
- Molekulargewicht:284.48 g/mol
- Merck-Index:13,08882
- Siedepunkt:361 °C (1013 hPa)
- Schmelzender Platingehalt:67 °C
- Dichte:0,94 g/cm³ (20 °C)
- Blitz Pt:196 °C (geschlossener Tiegel)
- Lagertemperatur:Kühlschrank
- EINECS:200-313-4
1 Varianten
Product Details & Documents
Synonyme: Octadecanoic acid, Octadecylic acid
- CAS Nummer:57-11-4
- Formel:C₁₈H₃₆O₂
- MDL-Nummer:MFCD00002752
- Molekulargewicht:284.48 g/mol
- Merck-Index:13,08882
- Siedepunkt:361 °C (1013 hPa)
- Schmelzender Platingehalt:67 °C
- Dichte:0,94 g/cm³ (20 °C)
- Blitz Pt:196 °C (geschlossener Tiegel)
- Lagertemperatur:Kühlschrank
- EINECS:200-313-4
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



