Methyloctanoat
Anbieter: MATREYA
Direkt zu
Über diesen Artikel
Synonyme: Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate
- CAS Nummer:111-11-5
- Formel:C₉H₁₈O₂
- MDL-Nummer:MFCD00009551
- Molekulargewicht:158.24 g/mol
- Siedepunkt:190 °C (1013 hPa)
- Schmelzender Platingehalt:–36,7 °C
- Dichte:0,877 g/cm³ (20 °C)
- Blitz Pt:68 °C
- EINECS:203-835-0
1 Varianten
Product Details & Documents
Synonyme: Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate
- CAS Nummer:111-11-5
- Formel:C₉H₁₈O₂
- MDL-Nummer:MFCD00009551
- Molekulargewicht:158.24 g/mol
- Siedepunkt:190 °C (1013 hPa)
- Schmelzender Platingehalt:–36,7 °C
- Dichte:0,877 g/cm³ (20 °C)
- Blitz Pt:68 °C
- EINECS:203-835-0
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



