Methyllaurat
Anbieter: MATREYA
Direkt zu
Über diesen Artikel
Synonyme: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester
- CAS Nummer:111-82-0
- Formel:C₁₃H₂₆O₂
- MDL-Nummer:MFCD00008966
- Molekulargewicht:214.35 g/mol
- Siedepunkt:266 °C (1013 hPa)
- Schmelzender Platingehalt:4,5 °C
- Dichte:0,8694 g/cm³ (20 °C)
- EINECS:203-911-3
1 Varianten
Product Details & Documents
Synonyme: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester
- CAS Nummer:111-82-0
- Formel:C₁₃H₂₆O₂
- MDL-Nummer:MFCD00008966
- Molekulargewicht:214.35 g/mol
- Siedepunkt:266 °C (1013 hPa)
- Schmelzender Platingehalt:4,5 °C
- Dichte:0,8694 g/cm³ (20 °C)
- EINECS:203-911-3



