Order Entry
Switzerland
ContactUsLinkComponent
Irritant
L(+)-Arginin
Catalog # BACE4030316.0250
Supplier:  BACHEM BIOCHEMICA
CAS Number:  
Irritant
L(+)-Arginin
Catalog # BACE4030316.0250
Supplier:  BACHEM BIOCHEMICA
CAS Number:  

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :174.2 g/mol
Merck Index:13,00785
Boiling Pt:368 °C (1013 hPa)
Melting Pt:238 °C
Density:1.3 g/cm³ (20 °C)
Storage Temperature:Raumtemperatur
EINECS:200-811-1
 :GHS07
CAS Number:74-79-3
Formula:C₆H₁₄N₄O₂
MDL Number:MFCD00002635