AuftragsverfolgungKontakt
Apoptosis inducer set
Apoptosis inducer set
Apoptosis inducer set, 1 SET
Catalog # ENZOALX850235KI01
Supplier:   Enzo Life Sciences
Apoptosis inducer set
Catalog # ENZOALX850235KI01
Supplier:  Enzo Life Sciences
CAS Number:  
Apoptosis inducer set
Apoptosis inducer set, 1 SET
CAS Number:  
Price
SFr. 751.00
Log in for availability
1 * 1 SET
Product Details & Documents
The set contains five reagents that induce apoptosis through different mechanisms.
  • Ready to use
Actinomycin D, an antineoplastic antibiotic, inhibits RNA synthesis. Camptothecin, an inhibitor of nuclear topoisomerase, induces apoptosis in many types of cells. Cycloheximide, an active antibiotic against many yeast and fungi, inhibits protein synthesis. Dexamethasone, an active and highly stable glucocorticoid, probably induces apoptosis by binding and activating the intracellular glucocorticoid receptor. Etoposide, a derivative of podophyllotoxin, inhibits topoisomerase activity.
Delivery information: Supplied with Actinomycin D, 50 µl (10 mM), violet cap, Camptothecin, 1 ml (2 mM), red cap, Cycloheximide, 1 ml (100 mM), green cap, Dexamethasone, 1 ml (10 mM), blue cap and Etoposide, 100 µl (100 mM), yellow cap.
Specifications
Specifications
  • VE:
    1 SET
  • Bezeichnung:
    Apoptosis inducer set
  • Cat. No.:
    ENZOALX850235KI01