ContactUsLinkComponent
Guanidiniumchlorid 8 mol/l in wässriger Lösung
Catalog # 786-279
Supplier:  G-Biosciences
CAS Number:  
undefined
Guanidiniumchlorid 8 mol/l in wässriger Lösung
Catalog # 786-279
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Packung:
    Glasflasche
  • VE:
    200 mL
  • Seq:
    2530
  • Environmentally Preferable:
Specifications
About this item
Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
 :
 :95.53 g/mol
EINECS:200-002-3
 :GHS07
Storage Temperature:Raumtemperatur
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026
Melting Pt:185
Merck Index:13,04578