DL-Homocystein
100 µl
Katalognr. CHMCAB5512
Anbieter: Chemicon
Direkt zu
Product Details & Documents
Synonyme: (±)-Homocysteine, 2-Amino-4-mercaptobutanoic acid, H-DL-HoCys-OH
- CAS Nummer:454-29-5
- Formel:C₄H₉NO₂S
- MDL-Nummer:MFCD00004898
- Molekulargewicht:135.19 g/mol
- Lagertemperatur:Gefrierschrank
- EINECS:207-222-9
Technische Daten
Technische Daten
- VE:100 µl
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



