SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standards
Organische Standards
β-Hexabromcyclododecan-[13C12] 50 µg/ml in Toluol (99% 13C)
β-Hexabromcyclododecan-[13C12] 50 µg/ml in Toluol (99% 13C)
CAMBCLM79231.2
:
CAMBRIDGE ISOTOPE LABS
:
β-Hexabromcyclododecan-[13C12] 50 µg/ml in Toluol (99% 13C)
CAMBCLM79231.2
:
CAMBRIDGE ISOTOPE LABS
:
Packung:
Glasflasche
VE:
1,2 mL
Bezeichnung:
β-Hexabromcyclododecan-[13C12] 50 µg/ml in Toluol (99% 13C)