Cleanroom wipes, Clino® Carbon
Katalognr. PPSA3600098
Anbieter: PFENNIG REINIGUNGSTECHNIK
Technische Daten
- Verpackt:5/pack
- Farbe:Weiß
- L×B:350×340 mm
- VE:5
Technische Daten
Über diesen Artikel
Reusable polyester/carbon cloth for use on all smooth and water-resistant surfaces.
- Low particle emission
- Electrostatically conductive
- Autoclavable
Suitable for use in ISO class 5 - 8 (GMP grade A/B, C, D) cleanrooms.