Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Agarose

Agarose

Marketsource
Supplier:   GERBU BIOCHEMICALS
Agarose
Supplier:  GERBU BIOCHEMICALS
Marketsource
Certificates
Agarose

4 Options Available Below

Product Details & Documents
  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Melting Pt:60…90 °C
  • Storage Temperature:Raumtemperatur
  • EINECS:232-731-8
Product Family Options

Product Information

  • Packung
    VE
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    200 g
    Log in for availability
    Log in for availability
    1 * 200 g
    1 * 200 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.