Human recombinant SENP2 (catalytic domain) (GST-tag) (from E. coli), GST Tag
Catalog # ENZOBMLUW97650100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- VE:100 µG
- Protein/Peptid-Typ:Rekombinant
- Ursprung:E. coli
- Spezies:Human
- Seq:02
- Tag sequence:GST Tag
- Biologische Aktivität:This fragment has full de-SUMOylating activity
- Protein/Peptid-Name:SENP2 (catalytic domain) (GST-tag)
Specifications
About this item
Produced in E. coli.