Human recombinant CREB binding protein (catalytic domain) (from E. coli)
ENZOBMLSE4520100
:
- VE:100 µG
- Protein/Peptid-Typ:Rekombinant
- Ursprung:E. coli
- Spezies:Human
- Seq:02
- Protein/Peptid-Name:CREB binding protein (catalytic domain)
- Purity:>65% (SDS-PAGE)
- Zusammensetzung:Liquid. In 50 mM TRIS-HCl, pH 8,0, containing 0,1 mM EDTA, 10% glycerol.
Produced in E. coli. Catalytic domain (aa 1319-1710) of human CBP.