CD14, soluble (human) ELISA Kit
ENZOALX850302KI01
:
- VE:1 KIT
- Reaktivität der primären Antikörper:Human
- Target protein:CD14
- Bezeichnung:CD14, soluble (human) ELISA kit
- Seq:02
- Empfindlichkeit:Range: 5 - 50 ng/ml
For the quantitative determination of soluble human CD14 in serum, plasma and cell culture medium.
: Contains 1 precoated ELISA module1 vial detecting antibody (POD-labelled monoclonal antibody to human CD14) (ready-to-use)1 vial human CD14 standard (recombinant human CD14, lyophilized)1 vial reference serum (2.7 ± 0.5, lyophilized)2 tablets PBS1 vial dilution buffer1 vial Tween 201 vial stopping solution (ready-to-use)1 vial substrate solution (ready-to-use).