AuftragsverfolgungKontakt
Acetyltransferase activity kit | Avantor
Acetyltransferase activity kit
Acetyltransferase activity kit, 1 KIT
Katalognr. ENZOADI907026
Anbieter:   Enzo Life Sciences
Acetyltransferase activity kit
Katalognr. ENZOADI907026
Anbieter:  Enzo Life Sciences
CAS Nummer:  
Acetyltransferase activity kit
Acetyltransferase activity kit, 1 KIT
CAS Nummer:  
Preis
SFr. 814.00
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
For the measurement of acetyl-CoA dependent acetyltransferase activity in purified in vitro samples that do not contain reductants from any species.
  • Universal - suitable for any AcetylCoA-dependent acetyltransferase
  • Robust - Z' factor = 0.87
  • Sensitive - fluorescent detection allows for both DNA and protein assays
  • Higher throughput - homogeneous mix-and-read assay
The Acetyltransferase activity kit is a homogeneous mix-and-read fluorescent assay for the determination of any acetyl-CoA dependent acetyltransferase activity. It is suitable for end-point or kinetic read options, which is ideal for determining mechanism of action, kinetics, and screening candidate compounds. The assay is amendable to HTS and miniaturization.
Bestellinformationen: Contains Black Microtiter plate, Reaction buffer, Assay buffer, Detection solution, Positive control.
Technische Daten
Technische Daten
  • VE:
    1 KIT
  • Bezeichnung:
    Acetyltransferase activity kit
Technische Daten