Anti-PRP Mouse Monoclonal Antibody [clone: 3F4]
Supplier: Enzo Life Sciences
Certificates
About this item
Recommended Applications: ELISA, IHC, IP, WB
Species reactivity: Human, Hamster, Cat
Type: Primary
Antigen: PRP
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Specifications
- Catalog No:
- ENZOENZABS1190200
- ENZOENZABS1190500
- ENZOENZABS1191000
- Antigen-Symbol:
- PRP
- PRP
- PRP
- Antigen-Name:
- Prion protein
- Prion protein
- Prion protein
- Antigen-Synonyme:
- CD230|PrP
- CD230|PrP
- CD230|PrP
- Antikörper-Typ:
- Primär
- Primär
- Primär
- Klonalität:
- Monoklonal
- Monoklonal
- Monoklonal
- Konjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Klon:
- 3F4
- 3F4
- 3F4
- Reaktivität:
- Hamster, Human, Cat
- Wirt:
- Mouse
- Mouse
- Mouse
- Isotyp:
- IgG2a
- IgG2a
- IgG2a
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- ImmunoChemie:
- Yes
- Yes
- Yes
- Immunpräzipitation:
- Yes
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Liquid
- Reinigung:
- Purified
- Purified
- Purified
- Lagerungspuffer:
- In PBS.
- In PBS.
- In PBS.
- Lagertemperatur:
- –20 °C
- –20 °C
- –20 °C
- Transporttemperatur:
- Shipped on blue ice
- Shipped on blue ice
- Shipped on blue ice