Anti-14-3-3 Rabbit Polyclonal Antibody
ENZOBMLSA4830100
- Antikörper-Typ:Primär
- Antigen-Name:14-3-3 protein
- Antigen-Symbol:14-3-3
- Klonalität:Polyklonal
- Wirt:Rabbit
- ImmunoChemie:Yes
- Immunpräzipitation:Yes
- Reaktivität:Mammals
- Western blot:Yes
- Seq:03
- Form:Liquid. Neat serum.
- Antigen-Synonyme:beta isoform|14-3-3 alpha|Protein 1054|Alpha polypeptide|Protein kinase C inhibitor protein 1|Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein|protein kinase C inhibitor protein-1|14-3-3 Protein beta/alpha|brain Protein 14-3-3
- VE:100 µl
Host: Rabbit
- For Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes mammalian, chicken, plants, yeast and invertebrate 14-3-3 pan specific.
Type: Primary
Antigen: 14-3-3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mammals