AuftragsverfolgungKontakt
Anti-Neuronal Calcium Sensor-1 Rabbit Polyclonal Antibody | Avantor
Anti-Neuronal Calcium Sensor-1 Rabbit Polyclonal Antibody
Katalognr.
Anti-Neuronal Calcium Sensor-1 Rabbit Polyclonal Antibody
Katalognr.
Anbieter:  
Anti-Neuronal Calcium Sensor-1 Rabbit Polyclonal Antibody
Product Details & Documents
Host: Rabbit
  • For Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes rat NCS-1.
Type: Primary
Antigen: NEURONAL CALCIUM SENSOR-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat