Order Entry
Switzerland
ContactUsLinkComponent
Anti-BCL2L2 Rat Monoclonal Antibody
Anti-BCL2L2 Rat Monoclonal Antibody
Catalog # ENZOALX804354C100
Supplier:  ENZO LIFE SCIENCES
Anti-BCL2L2 Rat Monoclonal Antibody
Catalog # ENZOALX804354C100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    BCL2 like 2
  • Antigen-Symbol:
    BCL2L2
  • Klonalität:
    Monoklonal
  • Wirt:
    Rat
  • Immunpräzipitation:
    Yes
  • Isotyp:
    IgG2a
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 25% glycerol and 0,02% sodium azide
  • Antigen-Synonyme:
    BCL2L2|KIAA0271|BCL-W|BCLW
  • Konzentration:
    1 mg/ml
  • Reinigung:
    Protein G-affinity purified
  • Größe:
    100 µg
  • VE:
    100 µG

Specifications

About this item

Host: Rat, Isotype: IgG2a

  • For Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, monkey and dog Bcl-w.

Type: Primary
Antigen: BCL2L2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human