Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # ENZOADISAB100J
Supplier: ENZO LIFE SCIENCES
Specifications
- Antikörper-Typ:Sekundär
- Antigen-Name:Immunoglobulin G
- Antigen-Symbol:IgG
- Klonalität:Polyklonal
- Konjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Wirt:Goat
- ImmunoChemie:Yes
- Western blot:Yes
- Seq:02
- Form:Liquid. In 0,005 M sodium phosphate, pH 7,6, containing 0,125 M sodium chloride, 7,5 mg/ml BSA (IgG-free, protease-free), and 50% glycerol.
- Reinigung:Immunoaffinity purified
- VE:1 mL
Specifications
About this item
Immunoglobulin G (IgG) is an immunoglobulin monomer consisting of two (gamma) heavy chains and two light chains. Each IgG molecule contains two antigen binding domains and a single effector (Fc) domain. Assay Designs offers isotyping kits and antibodies in a variety of formats for the detection of human, mouse, rabbit, rat, sheep, and goat IgG.
- For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Recognizes mouse IgG.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse