Reagent for sulphate determination using Lovibond® MD photometers
Catalog # LOVI515450BT
Supplier: Lovibond
CAS Number:
Specifications
- Chemische Methode:Barium sulphate (turbidity)
- VE:100 Tab.
- Bezeichnung:Sulphate tablet
- Für Gerät:MD600, MD610, MultiDirect and Spectrodirect
- Messbereich:5 - 100 mg/l SO₄